Assay Details
| Gene Symbol: IQCF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075773.1 | NP_001069241.1 | 1 - 2 | 88 | 96 | |
| GenBank mRNA | BC103417.1 | - | 1 - 2 | 88 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103417.1, NM_001075773.1 transcripts.
Set Membership: |
|

IQCF1
Genomic Map