Assay Details
| Gene Symbol: IQCF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075773.1 | NP_001069241.1 | 2 - 3 | 197 | 67 | |
| GenBank mRNA | BC103417.1 | - | 2 - 3 | 197 | 67 | |
More Information
Set Membership: |
|

IQCF1
Genomic Map