Assay Details
| Gene Symbol: COQ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076210.2 | NP_001069678.1 | 6 - 7 | 912 | 67 | |
| GenBank mRNA | BC102147.1 | - | 6 - 7 | 913 | 67 | |
More Information
Set Membership: |
|

COQ3
Genomic Map