Assay Details
| Gene Symbol: POLR2I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076329.2 | NP_001069797.1 | 72 | 92 | ||
| GenBank mRNA | BC108233.1 | - | 74 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLR2I| Gene Symbol: POLR2I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076329.2 | NP_001069797.1 | 72 | 92 | ||
| GenBank mRNA | BC108233.1 | - | 74 | 92 | ||
Set Membership: |
|