Assay Details
| Gene Symbol: BLOC1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076362.2 | NP_001069830.1 | 2 - 3 | 220 | 64 | |
| GenBank mRNA | BC111605.1 | - | 2 - 3 | 221 | 64 | |
More Information
Set Membership: |
|

BLOC1S1
Genomic Map