Assay Details
| Gene Symbol: MRPL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076514.2 | NP_001069982.1 | 5 - 6 | 470 | 68 | |
| GenBank mRNA | BC109734.1 | - | 5 - 6 | 471 | 68 | |
More Information
Set Membership: |
|

MRPL21
Genomic Map