Assay Details
| Gene Symbol: C5H12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076992.1 | NP_001070460.1 | 1 - 2 | 74 | 69 | |
| GenBank mRNA | BC102663.1 | - | 1 - 2 | 74 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102663.1, NM_001076992.1 transcripts.
Set Membership: |
|

C5H12orf54
Genomic Map