Assay Details
| Gene Symbol: C5H12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076992.1 | NP_001070460.1 | 2 - 3 | 190 | 73 | |
| GenBank mRNA | BC102663.1 | - | 2 - 3 | 190 | 73 | |
More Information
Set Membership: |
|

C5H12orf54
Genomic Map