Assay Details
| Gene Symbol: GML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077025.2 | NP_001070493.1 | 1 - 2 | 91 | 56 | |
| GenBank mRNA | BC108209.1 | - | 1 - 2 | 92 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC108209.1, NM_001077025.2 transcripts.
Set Membership: |
|

GML
Genomic Map