Assay Details
| Gene Symbol: MRPL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077884.1 | NP_001071352.1 | 4 - 5 | 398 | 67 | |
| GenBank mRNA | BC108192.1 | - | 4 - 5 | 398 | 67 | |
More Information
Set Membership: |
|

MRPL23
Genomic Map