Assay Details
| Gene Symbol: SLC25A20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077936.2 | NP_001071404.1 | 7 - 8 | 814 | 77 | |
| GenBank mRNA | BC103013.1 | - | 7 - 8 | 815 | 77 | |
More Information
Set Membership: |
|

SLC25A20
Genomic Map