Assay Details
| Gene Symbol: RPL36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078139.2 | NP_001071607.1 | 1 | 259 | 53 | |
| GenBank mRNA | BC102085.1 | - | 1 | 260 | 53 | |
| BC141994.1 | - | 2 - 3 | 419 | 53 | ||
More Information
Set Membership: |
|

RPL36
Genomic Map