Assay Details
| Gene Symbol: CDC26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078153.2 | NP_001071621.1 | 3 - 4 | 393 | 100 | |
| GenBank mRNA | BC102716.1 | - | 3 - 4 | 394 | 100 | |
More Information
Set Membership: |
|

CDC26
Genomic Map