Assay Details
| Gene Symbol: LDAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079608.2 | NP_001073076.1 | 3 - 4 | 385 | 75 | |
| GenBank mRNA | BC102736.1 | - | 3 - 4 | 386 | 75 | |
More Information
Set Membership: |
|

LDAH
Genomic Map