Assay Details
| Gene Symbol: CHRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080733.1 | NP_001074202.1 | 1 | 1142 | 62 | |
| GenBank mRNA | DQ154113.1 | - | 1 | 1142 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHRM2| Gene Symbol: CHRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080733.1 | NP_001074202.1 | 1 | 1142 | 62 | |
| GenBank mRNA | DQ154113.1 | - | 1 | 1142 | 62 | |
Set Membership: |
|