Assay Details
| Gene Symbol: CXCR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174301.3 | NP_776726.1 | 1 - 2 | 102 | 71 | |
| GenBank mRNA | AF399642.1 | - | 1 - 2 | 58 | 71 | |
| BC105217.1 | - | 1 - 2 | 102 | 71 | ||
| M86739.1 | - | 1 - 2 | 74 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105217.1, NM_174301.3 transcripts.
Set Membership: |
|

CXCR4
Genomic Map