Assay Details
| Gene Symbol: HADHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174336.3 | NP_776761.1 | 3 - 4 | 150 | 72 | |
| GenBank mRNA | AJ003066.1 | - | 3 - 4 | 234 | 72 | |
| BC102638.1 | - | 3 - 4 | 150 | 72 | ||
| BT030566.1 | - | 3 - 4 | 188 | 72 | ||
More Information
Set Membership: |
|

HADHB
Genomic Map