Assay Details
| Gene Symbol: LEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174559.4 | NP_776984.1 | 1 - 2 | 82 | 94 | |
| GenBank mRNA | BC108204.1 | - | 1 - 2 | 83 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC108204.1, NM_174559.4 transcripts.
Set Membership: |
|

LEAP2
Genomic Map