Assay Details
| Gene Symbol: LEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174559.4 | NP_776984.1 | 2 - 3 | 219 | 76 | |
| GenBank mRNA | AJ409014.1 | - | 2 - 3 | 194 | 76 | |
| BC108204.1 | - | 2 - 3 | 220 | 76 | ||
More Information
Set Membership: |
|

LEAP2
Genomic Map