Assay Details
| Gene Symbol: PYCARD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174730.2 | NP_777155.1 | 2 - 3 | 396 | 57 | |
| GenBank mRNA | AB050006.1 | - | 2 - 3 | 329 | 57 | |
| BC102385.1 | - | 2 - 3 | 396 | 57 | ||
More Information
Set Membership: |
|

PYCARD
Genomic Map