Assay Details
| Gene Symbol: GNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175777.3 | NP_786971.2 | 3 - 4 | 357 | 59 | |
| GenBank mRNA | BC105260.1 | - | 3 - 4 | 357 | 59 | |
| BT030728.1 | - | 2 - 3 | 91 | 59 | ||
| M13236.1 | - | 3 - 4 | 192 | 59 | ||
| X03073.1 | - | 236 | 59 | |||
More Information
Set Membership: |
|

GNB1
Genomic Map