Assay Details
| Gene Symbol: HEY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001172.3 | NP_001001172.2 | 4 - 5 | 454 | 88 | |
| GenBank mRNA | AB118750.1 | - | 4 - 5 | 472 | 88 | |
| BC112574.1 | - | 4 - 5 | 455 | 88 | ||
More Information
Set Membership: |
|

HEY1
Genomic Map