Assay Details
| Gene Symbol: FANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003904.3 | NP_001003904.2 | 5 - 6 | 580 | 60 | |
| GenBank mRNA | AY675079.1 | - | 5 - 6 | 490 | 60 | |
| BC109735.1 | - | 5 - 6 | 581 | 60 | ||
More Information
Set Membership: |
|

FANK1
Genomic Map