Assay Details
| Gene Symbol: BUD23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034457.2 | NP_001029629.2 | 2 - 3 | 120 | 70 | |
| GenBank mRNA | BC109889.1 | - | 2 - 3 | 120 | 70 | |
| BT021557.1 | - | 2 - 3 | 145 | 70 | ||
More Information
Set Membership: |
|

BUD23
Genomic Map