Assay Details
| Gene Symbol: NRM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037596.2 | NP_001032685.1 | 3 - 4 | 704 | 62 | |
| GenBank mRNA | BC109506.1 | - | 3 - 4 | 707 | 62 | |
More Information
Set Membership: |
|

NRM
Genomic Map