Assay Details
| Gene Symbol: ADTRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037621.1 | NP_001032710.1 | 4 - 5 | 731 | 71 | |
| GenBank mRNA | BC109489.1 | - | 4 - 5 | 731 | 71 | |
More Information
Set Membership: |
|

ADTRP
Genomic Map