Assay Details
| Gene Symbol: CAMLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037625.2 | NP_001032714.1 | 3 - 4 | 747 | 141 | |
| GenBank mRNA | BC109601.1 | - | 3 - 4 | 748 | 141 | |
More Information
Set Membership: |
|

CAMLG
Genomic Map