Assay Details
| Gene Symbol: UBE2J2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038043.1 | NP_001033132.1 | 2 - 3 | 155 | 72 | |
| GenBank mRNA | BC111185.1 | - | 3 - 4 | 187 | 72 | |
More Information
Set Membership: |
|

UBE2J2
Genomic Map