Assay Details
| Gene Symbol: AARSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038073.1 | NP_001033162.1 | 8 - 9 | 949 | 63 | |
| GenBank mRNA | BC109537.1 | - | 8 - 9 | 949 | 63 | |
More Information
Set Membership: |
|

AARSD1
Genomic Map