Assay Details
| Gene Symbol: RPP38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038085.2 | NP_001033174.1 | 2 - 3 | 247 | 133 | |
| GenBank mRNA | BC109652.1 | - | 2 - 3 | 248 | 133 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109652.1, NM_001038085.2 transcripts.
Set Membership: |
|

RPP38
Genomic Map