Assay Details
| Gene Symbol: CRISP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038089.2 | NP_001033178.1 | 7 - 8 | 619 | 116 | |
| GenBank mRNA | BC109478.1 | - | 7 - 8 | 620 | 116 | |
More Information
Set Membership: |
|

CRISP2
Genomic Map