Assay Details
| Gene Symbol: PRELID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038138.2 | NP_001033227.1 | 4 - 5 | 684 | 84 | |
| GenBank mRNA | BC110001.1 | - | 4 - 5 | 685 | 84 | |
More Information
Set Membership: |
|

PRELID1
Genomic Map