Assay Details
| Gene Symbol: ZNF330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038157.2 | NP_001033246.1 | 4 - 5 | 333 | 154 | |
| GenBank mRNA | BC109643.1 | - | 4 - 5 | 334 | 154 | |
More Information
Set Membership: |
|

ZNF330
Genomic Map