Assay Details
| Gene Symbol: NOA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038188.2 | NP_001033277.1 | 5 - 6 | 1921 | 82 | |
| GenBank mRNA | BC109654.1 | - | 5 - 6 | 1922 | 82 | |
More Information
Set Membership: |
|

NOA1A
Genomic Map