Assay Details
| Gene Symbol: ARGLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038190.2 | NP_001033279.1 | 2 - 3 | 802 | 77 | |
| GenBank mRNA | BC111128.1 | - | 2 - 3 | 803 | 77 | |
More Information
Set Membership: |
|

ARGLU1
Genomic Map