Assay Details
| Gene Symbol: CCT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038194.1 | NP_001033283.1 | 5 - 6 | 694 | 65 | |
| GenBank mRNA | BC111225.1 | - | 5 - 6 | 694 | 65 | |
More Information
Set Membership: |
|

CCT4
Genomic Map