Assay Details
| Gene Symbol: CMSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038215.2 | NP_001033304.1 | 2 - 3 | 271 | 75 | |
| GenBank mRNA | BC111214.1 | - | 2 - 3 | 272 | 75 | |
More Information
Set Membership: |
|

CMSS1
Genomic Map