Assay Details
| Gene Symbol: ATP5PB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038501.2 | NP_001033590.1 | 1 - 2 | 76 | 95 | |
| GenBank mRNA | BC110212.1 | - | 1 - 2 | 77 | 95 | |
More Information
Set Membership: |
|

ATP5PB
Genomic Map