Assay Details
| Gene Symbol: ATP5PB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038501.2 | NP_001033590.1 | 3 - 4 | 256 | 83 | |
| GenBank mRNA | BC110212.1 | - | 3 - 4 | 257 | 83 | |
| X06088.1 | - | 94 | 83 | |||
More Information
Set Membership: |
|

ATP5PB
Genomic Map