Assay Details
| Gene Symbol: CENPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038576.2 | NP_001033665.1 | 4 - 5 | 317 | 98 | |
| GenBank mRNA | BC109754.1 | - | 4 - 5 | 318 | 98 | |
More Information
Set Membership: |
|

CENPS
Genomic Map