Assay Details
| Gene Symbol: ETFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038582.1 | NP_001033671.1 | 1 - 2 | 90 | 66 | |
| GenBank mRNA | BC109603.1 | - | 1 - 2 | 90 | 66 | |
More Information
Set Membership: |
|

ETFB
Genomic Map