Assay Details
| Gene Symbol: XPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038681.2 | NP_001033770.1 | 4 - 5 | 579 | 121 | |
| GenBank mRNA | BC110270.1 | - | 4 - 5 | 580 | 121 | |
More Information
Set Membership: |
|

XPA
Genomic Map