Assay Details
| Gene Symbol: IGFBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040495.2 | NP_001035585.1 | 2 - 3 | 521 | 74 | |
| GenBank mRNA | BC110141.1 | - | 2 - 3 | 522 | 74 | |
| S52774.1 | - | 2 - 3 | 221 | 74 | ||
More Information
Set Membership: |
|

IGFBP6
Genomic Map