Assay Details
| Gene Symbol: GTF2H5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040537.2 | NP_001035627.1 | 1 - 2 | 23 | 65 | |
| GenBank mRNA | BC111195.1 | - | 1 - 2 | 25 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111195.1, NM_001040537.2 transcripts.
Set Membership: |
|

GTF2H5
Genomic Map