Assay Details
| Gene Symbol: MED27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040546.2 | NP_001035636.1 | 1 - 2 | 229 | 63 | |
| GenBank mRNA | BC109883.1 | - | 1 - 2 | 230 | 63 | |
More Information
Set Membership: |
|

MED27
Genomic Map