Assay Details
| Gene Symbol: DIABLO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045882.2 | NP_001039347.1 | 1 - 2 | 86 | 56 | |
| GenBank mRNA | BC111127.1 | - | 1 - 2 | 87 | 56 | |
More Information
Set Membership: |
|

DIABLO
Genomic Map