Assay Details
| Gene Symbol: RNASEH2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045921.1 | NP_001039386.1 | 3 - 4 | 514 | 87 | |
| GenBank mRNA | BC111611.1 | - | 3 - 4 | 514 | 87 | |
More Information
Set Membership: |
|

RNASEH2C
Genomic Map