Assay Details
| Gene Symbol: SMDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045937.1 | NP_001039402.1 | 2 - 3 | 358 | 116 | |
| GenBank mRNA | BC111682.1 | - | 2 - 3 | 358 | 116 | |
More Information
Set Membership: |
|

SMDT1
Genomic Map