Assay Details
| Gene Symbol: GCDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045939.2 | NP_001039404.1 | 6 - 7 | 598 | 70 | |
| GenBank mRNA | BC112822.1 | - | 6 - 7 | 599 | 70 | |
More Information
Set Membership: |
|

GCDH
Genomic Map