Assay Details
| Gene Symbol: RCAN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045945.1 | NP_001039410.1 | 4 - 5 | 689 | 62 | |
| GenBank mRNA | BC112714.1 | - | 4 - 5 | 689 | 62 | |
More Information
Set Membership: |
|

RCAN3
Genomic Map